Novus Biologicals
Manufacturer Code:NBP294034100UL
Catalog # NB022632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human TCEB2 (NP_009039.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18-kD subunit EloB Elongin 18 kDa subunit elongin-B RNA polymerase II transcription factor SIII p18 subunit RNA polymerase II transcription factor SIII subunit B SIII p18 transcription elongation factor B (SIII) polypeptide 2 (18kD elongin B) transcription elongation factor B (SIII) polypeptide 2 (18kDa elongin B) transcription elongation factor B polypeptide 2; EloB; Elongin 18 kDa subunit; elongin B; elongin, 18-kD subunit; Elongin-B; RNA polymerase II transcription factor SIII p18 subunit; RNA polymerase II transcription factor SIII subunit B; SIII p18; transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B); Transcription elongation factor B polypeptide 2
Gene Aliases: ELOB; SIII; TCEB2
UniProt ID: (Human) Q15370
Entrez Gene ID: (Human) 6923
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.