Novus Biologicals
Manufacturer Code:NBP246085
Catalog # NBP246085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G10 |
Immunogen | Full length human recombinant protein of human TCEAL1 (NP_004771) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Nuclear phosphoprotein p21/SIIR; Nuclear phosphoprotein p21/SIIR p21 pp21 SIIRTCEA-like protein 1 transcription elongation factor A (SII)-like 1 transcription elongation factor A protein-like 1 Transcription elongation factor S-II protein-like 1; TCEA-like protein 1; transcription elongation factor A (SII)-like 1; Transcription elongation factor A protein-like 1; Transcription elongation factor S-II protein-like 1
Gene Aliases: p21; pp21; SIIR; TCEAL1; WEX9
UniProt ID: (Human) Q15170
Entrez Gene ID: (Human) 9338
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.