OriGene
Manufacturer Code:CF808419
Catalog # 50-166-8841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI11H10 |
Immunogen | Full length human recombit protein of human TCAP produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 19 kDa sarcomeric protein; CMD1N CMH25 LGMD2G T-cap TELE telethonin; limb girdle muscular dystrophy 2G (autosomal recessive); Telethonin; teneurin C-terminal associated peptide; Titin cap protein
Gene Aliases: CMD1N; CMH25; LGMD2G; T-cap; TCAP; TELE; telethonin
UniProt ID: (Human) O15273
Entrez Gene ID: (Human) 8557
Molecular Function:
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.