R&D Systems
Manufacturer Code:MAB1930
Catalog # MAB1930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 252294 |
Immunogen | <EM>E. coli</EM>-derived recombinant human TC-PTP Pro2-Asn314 Accession # P17706 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: protein tyrosine phosphatase non-receptor type 2 PTP2 PTPN2 PTPTPTN2 T-cell protein tyrosine phosphatase T-cell protein-tyrosine phosphatase TCELLPTP TCPTP TC-PTP TCPTPEC 3.1.3.48 tyrosine-protein phosphatase non-receptor type 2; T-cell protein tyrosine phosphatase; T-cell protein-tyrosine phosphatase; TCPTP; Tyrosine-protein phosphatase non-receptor type 2
Gene Aliases: PTN2; PTPN2; PTPT; TC-PTP; TCELLPTP; TCPTP
UniProt ID: (Human) P17706
Entrez Gene ID: (Human) 5771
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.