Novus Biologicals
Manufacturer Code:NBP290722AF532
Catalog # NB076924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse TC-PTP/PTPN2 (Q06180-1 Ser2-Asn314). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: protein tyrosine phosphatase non-receptor type 2 PTPTPTN2 T-cell protein tyrosine phosphatase T-cell protein-tyrosine phosphatase TCELLPTP TC-PTP TCPTPEC 3.1.3.48 tyrosine-protein phosphatase non-receptor type 2; T-cell protein tyrosine phosphatase; T-cell protein-tyrosine phosphatase; TCPTP; Tyrosine-protein phosphatase non-receptor type 2
Gene Aliases: PTN2; PTPN2; PTPT; TC-PTP; TCELLPTP; TCPTP
UniProt ID: (Human) P17706
Entrez Gene ID: (Human) 5771
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.