Novus Biologicals
Manufacturer Code:NBP29372720UL
Catalog # NB029318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-106 of human C8orf4 (NP_064515.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 8 open reading frame 4 hTC-1 MGC22806 TC1 thyroid cancer-1 TC-1human thyroid cancer 1 Thyroid cancer protein 1; human thyroid cancer 1; TC-1; Thyroid cancer protein 1; Transcriptional and immune response regulator
Gene Aliases: C8orf4; TC-1; TC1; TCIM
UniProt ID: (Human) Q9NR00
Entrez Gene ID: (Human) 56892
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.