OriGene
Manufacturer Code:TA700230
Catalog # 50-167-7746
Quantity:
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C1 |
Immunogen | Full length human recombit protein of human TBXAS1 produced in HEK293T cell. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BDPLT14 CYP5 CYP5A1 GHOSAL THAS TS TXAS TXS; Cytochrome P450 5A1; cytochrome P450, family 5, subfamily A, polypeptide 1; Hydroperoxy icosatetraenoate dehydratase; platelet, cytochrome P450, subfamily V; thromboxane A synthase 1 (platelet); Thromboxane-A synthase; TXA synthase
Gene Aliases: BDPLT14; CYP5; CYP5A1; GHOSAL; TBXAS1; THAS; TS; TXAS; TXS
UniProt ID: (Human) P24557
Entrez Gene ID: (Human) 6916
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.