Invitrogen
Manufacturer Code:PA568616
Catalog # PIPA568616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat TBX4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SPS T-box protein 4 T-box transcription factor TBX4; T-box 4; T-box 4 protein; T-box protein 4; T-box transcription factor TBX4; transcription factor Tbx4
Gene Aliases: 3930401C23; ICPPS; SPS; TBX4
UniProt ID: (Human) P57082, (Dog) Q861Q9, (Mouse) P70325
Entrez Gene ID: (Bovine) 508750, (Pig) 100620126, (Rabbit) 100350522, (Human) 9496, (Dog) 403514, (Mouse) 21387, (Rat) 303399
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.