OriGene
Manufacturer Code:CF811209
Catalog # 50-167-0524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8G5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 244-481 of human TBX3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bladder cancer related protein XHL; T-box protein 3; T-box transcription factor TBX3; TBX3-ISO UMS XHL
Gene Aliases: TBX3; TBX3-ISO; UMS; XHL
UniProt ID: (Human) O15119
Entrez Gene ID: (Human) 6926
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.