Proteintech
Manufacturer Code:137001AP150UL
Catalog # 50-172-8943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBX21T-bet Fusion Protein Ag4618 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T bet T box 21 T box protein 21 T PET TBET TBLYM TBX21 TBX21T bet Transcription factor TBLYM; T-box expressed in T cells; T-box protein 21; T-box transcription factor TBX21; T-cell-specific T-box transcription factor T-bet; Transcription factor TBLYM
Gene Aliases: T-bet; T-PET; TBET; TBLYM; TBT1; TBX21
UniProt ID: (Human) Q9UL17, (Mouse) Q9JKD8
Entrez Gene ID: (Human) 30009, (Mouse) 57765
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.