R&D Systems
Manufacturer Code:MAB8124SP
Catalog # MAB8124SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 668710 |
Immunogen | <EM>E. coli</EM>-derived recombinant human TBX20 Met1-Leu116 Accession # Q9UMR3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ASD4 T-box 20 T-box protein 20 T-box transcription factor TBX20; T-box protein 20; T-box transcription factor TBX20
Gene Aliases: ASD4; TBX20
UniProt ID: (Human) Q9UMR3
Entrez Gene ID: (Human) 57057
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.