Invitrogen
Manufacturer Code:PA5110464
Catalog # PIPA5110464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human TBX20 (NP_001071121.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430010M06Rik AL022859 ASD4 BOS_4292 cb826 H15-related T-box transcription factor hrT hrt T-box protein 20 T-box transcription factor TBX20 Tbx12 TBX20 whz ZF-TBX20; T-box protein 20; T-box transcription factor TBX20
Gene Aliases: ASD4; TBX20
UniProt ID: (Human) Q9UMR3
Entrez Gene ID: (Human) 57057, (Rat) 315497
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.