R&D Systems
Manufacturer Code:MAB5040
Catalog # MAB5040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 393725 |
Immunogen | <EM>E. coli</EM>-derived recombinant human TBX2 Thr593-Arg702 Accession # Q13207 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ10169 T-box 2 T-box protein 2 T-box transcription factor TBX2; T-box protein 2; T-box transcription factor TBX2
Gene Aliases: TBX2
UniProt ID: (Human) Q13207
Entrez Gene ID: (Human) 6909
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.