Invitrogen
Manufacturer Code:PA5116010
Catalog # PIPA5116010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TBX 2/3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bladder cancer related protein XHL; T-box 3 (ulnar mammary syndrome); T-box protein 2; T-box protein 3; T-box transcription factor TBX2; T-box transcription factor TBX3; TBX2 TBX3 TBX3/2
Gene Aliases: D5Ertd189e; TBX2; TBX3; TBX3-ISO; UMS; XHL
UniProt ID: (Human) Q13207, (Human) O15119, (Mouse) Q60707, (Mouse) P70324, (Rat) Q7TST9
Entrez Gene ID: (Human) 6909, (Human) 6926, (Mouse) 21385, (Rat) 303398, (Mouse) 21386, (Rat) 353305
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.