Proteintech
Manufacturer Code:232371AP150UL
Catalog # 50-173-4025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBX18 Fusion Protein Ag19258 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T box 18 T box protein 18 TBX18; T-box protein 18; T-box transcription factor TBX18
Gene Aliases: 2810012F10Rik; 2810404D13Rik; CAKUT2; TBX18
UniProt ID: (Human) O95935, (Mouse) Q9EPZ6
Entrez Gene ID: (Human) 9096, (Rat) 315870, (Mouse) 76365
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.