Novus Biologicals
Manufacturer Code:NBP274470AF594
Catalog # NP274470A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C2 |
Immunogen | Full length recombinant protein of human TBX1 (NP_542377) produced in HEK293T cells. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: brachyury; brachyury CAFS CTHM DGCR DGS DORV T-box 1 T-box 1 transcription factor C T-box protein 1 T-box transcription factor TBX1 TBX1C Testis-specific T-box protein TGA VCFS; T-box 1 transcription factor C; T-box protein 1; T-box transcription factor TBX1; Testis-specific T-box protein
Gene Aliases: CAFS; CATCH22; CTHM; DGCR; DGS; DORV; TBX1; TBX1C; TGA; VCF; VCFS
UniProt ID: (Human) O43435
Entrez Gene ID: (Human) 6899
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.