OriGene
Manufacturer Code:TA507326
Catalog # 50-167-7063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C2 |
Immunogen | Full length human recombit protein of human TBX1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: brachyury; CAFS CATCH22 CTHM DGCR DGS DORV TBX1C TGA VCF VCFS; T-box 1 transcription factor C; T-box protein 1; T-box transcription factor TBX1; Testis-specific T-box protein
Gene Aliases: CAFS; CATCH22; CTHM; DGCR; DGS; DORV; TBX1; TBX1C; TGA; VCF; VCFS
UniProt ID: (Human) O43435
Entrez Gene ID: (Human) 6899
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.