Invitrogen
Manufacturer Code:PA540712
Catalog # PIPA540712
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human TBX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brachyury; T-box 1; T-box 1 DGS TGA CAFS CTHM DGCR DORV VCFS TBX1C; T-box 1 transcription factor C; T-box protein 1; T-box transcription factor TBX1; Testis-specific T-box protein
Gene Aliases: CAFS; CATCH22; CTHM; DGCR; DGS; DORV; TBX1; TBX1C; TGA; VCF; VCFS
UniProt ID: (Human) O43435, (Mouse) P70323
Entrez Gene ID: (Rabbit) 100355164, (Human) 6899, (Dog) 608214, (Guinea pig) 100729688, (Mouse) 21380, (Rat) 360737
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.