Proteintech
Manufacturer Code:162451AP150UL
Catalog # 50-173-0824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBRG4 Fusion Protein Ag9176 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell cycle progression 2 protein; Cell cycle progression protein 2; Cell cycle progression restoration protein 2; CPR2 FASTKD4 KIAA0948 Protein TBRG4 TBRG4; FAST kinase domain-containing protein 4; FAST kinase domains 4; H_TD2522F11.8; Protein TBRG4; transforming growth factor beta regulated gene 4; Transforming growth factor beta regulator 4
Gene Aliases: 2310042P22Rik; AA120735; AA408001; AI527316; CPR2; FASTKD4; KIAA0948; mKIAA0948; R74877; Tb-12; TBRG4
UniProt ID: (Human) Q969Z0, (Mouse) Q91YM4, (Rat) Q5M9G9
Entrez Gene ID: (Human) 9238, (Mouse) 21379, (Rat) 360977
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.