Invitrogen
Manufacturer Code:PA513390
Catalog # PIPA513390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 6-36 amino acids from the N-terminal region of human TBRG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nuclear interactor of ARF and Mdm2; Transforming growth factor beta regulator 1; Transforming growth factor beta regulator 1 Nuclear interactor of ARF and Mdm2 TBRG1 NIAM
Gene Aliases: AA408552; AA409675; mFLJ00213; NIAM; TB-5; TBRG1
UniProt ID: (Human) Q3YBR2, (Mouse) Q3UB74
Entrez Gene ID: (Human) 84897, (Mouse) 21376
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.