Proteintech
Manufacturer Code:133461AP150UL
Catalog # 50-172-8669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBRG1 Fusion Protein Ag4179 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NIAM TB 5 TBRG1; Nuclear interactor of ARF and Mdm2; Transforming growth factor beta regulator 1
Gene Aliases: NIAM; TB-5; TBRG1
UniProt ID: (Human) Q3YBR2
Entrez Gene ID: (Human) 84897
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.