Invitrogen
Manufacturer Code:MA537916
Catalog # PIMA537916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2198 |
Immunogen | A synthesized peptide derived from human TBR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: T-box brain gene 1; T-box brain gene 1 T-box brain protein 1 T-box brain transcription factor 1 T-box brain 1 T-box brain 1 T-box brain 1-like Tbr1 TBR-1 T-brain-1 TES-56; T-box brain protein 1; T-box, brain, 1; T-box, brain, 1-like; T-brain-1; TBR-1; TES-56
Gene Aliases: TBR-1; TBR1; TES-56
UniProt ID: (Human) Q16650, (Mouse) Q64336
Entrez Gene ID: (Human) 10716, (Mouse) 21375, (Rat) 680427
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.