Invitrogen
Manufacturer Code:PA568787
Catalog # PIPA568787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the n terminal region of mouse TBR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T-box brain gene 1; T-box brain protein 1; T-box, brain 1; T-box, brain, 1; T-box, brain, 1-like; T-brain-1; TBR-1; TBR-1 TES-56 T-box brain protein 1 T-brain-1; TES-56
Gene Aliases: TBR-1; TBR1; TES-56
UniProt ID: (Human) Q16650, (Mouse) Q64336
Entrez Gene ID: (Human) 10716, (Dog) 488370, (Bovine) 505142, (Rabbit) 100340143, (Horse) 100061032, (Mouse) 21375, (Rat) 680427, (Guinea pig) 100717884
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.