Invitrogen
Manufacturer Code:PA534582
Catalog # PIPA534582
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 50 - 150 of Mouse TBR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T-box brain gene 1; T-box brain protein 1; T-box, brain, 1; T-box, brain, 1-like; T-brain-1; TBR-1; TBR-1 TES-56 T-box brain protein 1 T-brain-1; TES-56
Gene Aliases: TBR-1; TBR1; TES-56
UniProt ID: (Human) Q16650, (Mouse) Q64336
Entrez Gene ID: (Human) 10716, (Mouse) 21375, (Rat) 680427
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.