Invitrogen
Manufacturer Code:PA5117379
Catalog # PIPA5117379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of mouse TBR1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: T-box brain gene 1; T-box brain gene 1 T-box brain protein 1 T-box brain transcription factor 1 T-box brain 1 T-box brain 1 T-box brain 1-like Tbr1 TBR-1 T-brain-1 TES-56; T-box brain protein 1; T-box, brain, 1-like; T-brain-1; TBR-1; TES-56
Gene Aliases: Tbr1
UniProt ID: (Mouse) Q64336
Entrez Gene ID: (Mouse) 21375, (Rat) 680427
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.