Invitrogen
Manufacturer Code:PA584876
Catalog # PIPA584876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TBPL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa TBP-like protein; 21-kDA TBP-like protein; Second TBP of unique DNA protein; STUD; STUD TLF TLP TRF2; TATA box-binding protein-like 1; TATA box-binding protein-related factor 2; TBP-like 1; TBP-like factor; TBP-related factor 2; TBP-related protein
Gene Aliases: MGC:8389; MGC:9620; STUD; TBPL1; TLF; TLP; TLP21; TRF2; TRP
UniProt ID: (Human) P62380
Entrez Gene ID: (Human) 9519
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.