Invitrogen
Manufacturer Code:PA541110
Catalog # PIPA541110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TBPL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa TBP-like protein; 21-kDA TBP-like protein; Second TBP of unique DNA protein; STUD; TATA box binding protein-like 1; TATA box binding protein-like protein; TATA box-binding protein related factor 2; TATA box-binding protein-like 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1-like protein; TATA box-binding protein-related factor 2; TBP-like 1; TBP-like 1 TLF TLP STUD TRF2 MGC:8389 MGC:9620; TBP-like factor; TBP-like protein 1; TBP-related factor 2; TBP-related protein
Gene Aliases: 4732475G08; AW011832; AW491032; D18347; MGC:8389; MGC:9620; STD; STUD; TBPL1; TLF; TLP; TLP21; trf; TRF2; TRP; zgc:92340
UniProt ID: (Human) P62380, (Bovine) Q32LB1, (Mouse) P62340, (Zebrafish) Q5U385
Entrez Gene ID: (Human) 9519, (Horse) 100067729, (Dog) 476209, (Bovine) 613818, (Rabbit) 100346981, (Rat) 689030, (Mouse) 237336, (Guinea pig) 100719408, (Zebrafish) 403035
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.