Abnova
Manufacturer Code:H00009519A01
Catalog # 89-106-095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. &bsim 91 a.a) partial recombinant protein with GST tag.,TBPL1 (NP_004856 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant TBPL1.
For Research Use Only
Protein Aliases: 21 kDa TBP-like protein; 21-kDA TBP-like protein; MGC:8389; MGC:9620; Second TBP of unique DNA protein; STUD; TATA box-binding protein-like 1; TATA box-binding protein-related factor 2; TBP-like 1; TBP-like factor; TBP-related factor 2; TBP-related protein; TLF; TLP; TRF2
Gene Aliases: MGC:8389; MGC:9620; STUD; TBPL1; TLF; TLP; TLP21; TRF2; TRP
UniProt ID: (Human) P62380
Entrez Gene ID: (Human) 9519
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.