Proteintech
Manufacturer Code:122581AP150UL
Catalog # 50-172-7838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBPL1 Fusion Protein Ag2880 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa TBP like protein STUD TBP like 1 TBP like factor TBP like protein 1 TBP related factor 2 TBP related protein TBPL1 TLF TLP TLP21 TRF2 TRP; 21 kDa TBP-like protein; 21-kDA TBP-like protein; Second TBP of unique DNA protein; STUD; TATA box binding protein-like 1; TATA box binding protein-like protein; TATA box-binding protein-like 1; TATA box-binding protein-like protein 1; TATA box-binding protein-related factor 2; TBP-like 1; TBP-like factor; TBP-like protein 1; TBP-related factor 2; TBP-related protein
Gene Aliases: 4732475G08; AW011832; AW491032; D18347; MGC:8389; MGC:9620; STD; STUD; TBPL1; TLF; TLP; TLP21; TRF2; TRP
UniProt ID: (Human) P62380, (Mouse) P62340
Entrez Gene ID: (Human) 9519, (Rat) 689030, (Mouse) 237336
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.