Invitrogen
Manufacturer Code:PA530861
Catalog # PIPA530861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 18 and 257 of Human TBLR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C21 DC42 IRA1 TBLR1 F-box-like/WD repeat-containing protein TBL1XR1 TBL1-related protein 1 nuclear receptor co-repressor/HDAC3 complex subunit nuclear receptor corepressor/HDAC3 complex subunit TBLR1 transducin beta-like 1X-related protein 1; F-box-like/WD repeat-containing protein TBL1XR1; nuclear receptor co-repressor/HDAC3 complex subunit; Nuclear receptor corepressor/HDAC3 complex subunit TBLR1; TBL1-related protein 1; Transducin beta-like 1X-related protein 1
Gene Aliases: C21; DC42; IRA1; MRD41; TBL1XR1; TBLR1
UniProt ID: (Human) Q9BZK7
Entrez Gene ID: (Human) 79718
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.