Invitrogen
Manufacturer Code:PA5116008
Catalog # PIPA5116008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TBLR1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8030499H02Rik A630076E03Rik AW539987 C21 C230089I12Rik DC42 F-box-like/WD repeat protein TBL1XR1 F-box-like/WD repeat-containing protein TBL1XR1 FLJ12894 IRA1 MRD41 nuclear receptor co-repressor/HDAC3 complex subunit nuclear receptor corepressor/HDAC3 complex subunit TBLR1 RGD1560053 TBL1-related protein 1 TBL1XR1 Tblr1 transducin (beta)-like 1 X-linked receptor 1 transducin (beta)-like 1X-linked receptor 1 transducin beta-like 1X-related protein 1; F-box-like/WD repeat protein TBL1XR1; F-box-like/WD repeat-containing protein TBL1XR1; nuclear receptor co-repressor/HDAC3 complex subunit; Nuclear receptor corepressor/HDAC3 complex subunit TBLR1; TBL1-related protein 1; Transducin beta-like 1X-related protein 1
Gene Aliases: 8030499H02Rik; A630076E03Rik; AW539987; C21; C230089I12Rik; DC42; IRA1; MRD41; RGD1560053; TBL1XR1; TBLR1
UniProt ID: (Human) Q9BZK7, (Mouse) Q8BHJ5
Entrez Gene ID: (Human) 79718, (Rat) 365755, (Mouse) 81004
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.