Novus Biologicals
Manufacturer Code:NB600270AF750
Catalog # NB60270A750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide that maps to a region between residues 100 and 150 of human TBL1-related Protein 1 using the numbering given in SwissProt entry Q9BZK7 (GeneID 79718). |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: C21 DC42 F-box-like/WD repeat-containing protein TBL1XR1 FLJ12894 IRA1Transducin beta-like 1X-related protein 1 nuclear receptor co-repressor/HDAC3 complex subunit Nuclear receptor corepressor/HDAC3 complex subunit TBLR1 TBLR1TBL1-related protein 1 transducin (beta)-like 1 X-linked receptor 1 transducin (beta)-like 1X-linked receptor 1; F-box-like/WD repeat-containing protein TBL1XR1; nuclear receptor co-repressor/HDAC3 complex subunit; Nuclear receptor corepressor/HDAC3 complex subunit TBLR1; TBL1-related protein 1; Transducin beta-like 1X-related protein 1
Gene Aliases: C21; DC42; IRA1; MRD41; TBL1XR1; TBLR1
UniProt ID: (Human) Q9BZK7
Entrez Gene ID: (Human) 79718
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.