Proteintech
Manufacturer Code:124881AP150UL
Catalog # 50-172-8018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBL2 Fusion Protein Ag3200 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TBL2 transducin (beta) like 2 Transducin beta like protein 2 WBSCR13 WS betaTRP; transducin (beta)-like 2; transducin -like 2; Transducin beta-like protein 2; Williams-Beuren syndrome chromosomal region 13 protein; Williams-Beuren syndrome chromosome region 13; WS beta-transducin repeats protein; WS-betaTRP
Gene Aliases: C76179; TBL2; UNQ563/PRO1125; WBSCR13; WS-betaTRP; WS-bTRP
UniProt ID: (Human) Q9Y4P3, (Mouse) Q9R099
Entrez Gene ID: (Human) 26608, (Mouse) 27368, (Rat) 686610
Molecular Function:
RNA binding protein
enzyme modulator
esterase
hydrolase
kinase inhibitor
kinase modulator
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.