Invitrogen
Manufacturer Code:PA5110461
Catalog # PIPA5110461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 328-527 of human TBCE (NP_003184.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610206D02Rik BOS_24010 C530005D02Rik HRD KCS KCS1 pac2 pmn TBCE Tubulin-folding cofactor E Tubulin-specific chaperone E zgc:123075; hypoparathyroidism, growth and mental retardation, and dysmorphism; Tubulin-folding cofactor E; Tubulin-specific chaperone E
Gene Aliases: HRD; KCS; KCS1; pac2; TBCE
UniProt ID: (Human) Q15813
Entrez Gene ID: (Human) 6905
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.