Proteintech
Manufacturer Code:120211AP150UL
Catalog # 50-172-7649
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBCE Fusion Protein Ag2650 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HRD KCS KCS1 pac2 TBCE tubulin folding cofactor E Tubulin specific chaperone E; hypoparathyroidism, growth and mental retardation, and dysmorphism; Tubulin-folding cofactor E; Tubulin-specific chaperone E
Gene Aliases: 2610206D02Rik; C530005D02Rik; HRD; KCS; KCS1; pac2; pmn; TBCE
UniProt ID: (Human) Q15813, (Rat) Q5FVQ9, (Mouse) Q8CIV8
Entrez Gene ID: (Human) 6905, (Rat) 361255, (Mouse) 70430
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.