Invitrogen
Manufacturer Code:PA524074
Catalog # PIPA524074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 491-520 amino acids from the C-terminal region of human TBCCD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TBCC domain-containing protein 1
Gene Aliases: 5730478M09Rik; AU015083; BB165076; TBCCD1
UniProt ID: (Human) Q9NVR7, (Mouse) Q640P7
Entrez Gene ID: (Human) 55171, (Mouse) 70573
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.