Invitrogen
Manufacturer Code:PA521710
Catalog # PIPA521710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 52 and 306 of TBCC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CFC; CFC tubulin-folding cofactor C tubulin-specific chaperone C; Tubulin-folding cofactor C; Tubulin-specific chaperone C
Gene Aliases: 2810055C19Rik; CFC; TBCC
UniProt ID: (Human) Q15814, (Mouse) Q8VCN9
Entrez Gene ID: (Human) 6903, (Mouse) 72726
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.