Novus Biologicals
Manufacturer Code:NBP294247100UL
Catalog # NB023050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-108 of human TBCA (NP_004598.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CFA; CFA chaperonin cofactor A TCP1-chaperonin cofactor A tubulin folding cofactor A Tubulin-folding cofactor A tubulin-specific chaperone a; chaperonin cofactor A; TCP1-chaperonin cofactor A; Tubulin-folding cofactor A; Tubulin-specific chaperone A
Gene Aliases: TBCA
UniProt ID: (Human) O75347
Entrez Gene ID: (Human) 6902
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.