Proteintech
Manufacturer Code:123041AP150UL
Catalog # 50-172-7877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBCA Fusion Protein Ag2950 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CFA; CFA TBCA TCP1 chaperonin cofactor A tubulin folding cofactor A Tubulin specific chaperone A; chaperonin cofactor A; co-chaperonin associated with a- & b-tubulin; cofactor A; TCP1-chaperonin cofactor A; tubulin cofactor a; Tubulin-folding cofactor A; Tubulin-specific chaperone A
Gene Aliases: TBCA; Tbca13
UniProt ID: (Human) O75347, (Mouse) P48428, (Rat) Q6PEC1
Entrez Gene ID: (Human) 6902, (Mouse) 21371, (Rat) 366995
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.