Proteintech
Manufacturer Code:170781AP150UL
Catalog # 50-173-1371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBC1D5 Fusion Protein Ag10991 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0210 TBC1 domain family member 5 TBC1 domain family member 5 TBC1D5; TBC1 domain family member 5; TBC1 domain family, member 5
Gene Aliases: 1600014N05Rik; KIAA0210; RGD1561626; TBC1D5
UniProt ID: (Human) Q92609, (Mouse) Q80XQ2
Entrez Gene ID: (Human) 9779, (Mouse) 72238, (Rat) 501088
Molecular Function:
G-protein modulator
enzyme modulator
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.