OriGene
Manufacturer Code:TA502695
Catalog # 50-167-3731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E6 |
Immunogen | Full length human recombit protein of human TBC1D4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Akt substrate of 160 kDa; AS160; AS160 NIDDM5; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC1 domain family member 4; TBC1 domain family, member 4
Gene Aliases: AS160; KIAA0603; NIDDM5; TBC1D4
UniProt ID: (Human) O60343
Entrez Gene ID: (Human) 9882
Molecular Function: G-protein modulator enzyme modulator hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.