Invitrogen
Manufacturer Code:PA570846
Catalog # PIPA570846
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse TBC1D23 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCV non-structural protein 4A-transactivated protein 1; HCV nonstructural protein 4A-transactivated protein 1; NS4ATP1 HCV non-structural protein 4A-transactivated protein 1 HCV nonstructural protein 4A-transactivated protein 1 TBC1 domain family member 23; TBC1 domain family member 23; TBC1 domain family, member 23
Gene Aliases: 4930451A13Rik; AU015720; AU043671; AU043778; D030022P07Rik; NS4ATP1; RGD1307925; TBC1D23; W51689; zgc:63756
UniProt ID: (Human) Q9NUY8, (Mouse) Q8K0F1, (Zebrafish) Q7SXV1
Entrez Gene ID: (Rabbit) 100352783, (Dog) 478539, (Human) 55773, (Horse) 100072293, (Mouse) 67581, (Guinea pig) 100722256, (Rat) 304019, (Zebrafish) 798995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.