Proteintech
Manufacturer Code:170021AP150UL
Catalog # 50-173-1321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TBC1D23 Fusion Protein Ag10697 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCV non-structural protein 4A-transactivated protein 1; HCV nonstructural protein 4A-transactivated protein 1; NS4ATP1 TBC1 domain family member 23 TBC1 domain family member 23 TBC1D23; TBC1 domain family member 23; TBC1 domain family, member 23
Gene Aliases: 4930451A13Rik; AU015720; AU043671; AU043778; D030022P07Rik; NS4ATP1; RGD1307925; TBC1D23; W51689
UniProt ID: (Human) Q9NUY8, (Mouse) Q8K0F1
Entrez Gene ID: (Human) 55773, (Mouse) 67581, (Rat) 304019
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.