Invitrogen
Manufacturer Code:PA5114201
Catalog # PIPA5114201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: ERTAASTFKD FELASAQQRP DMVLRQRFRG LLRPEDRTKD VLTKPRTNRF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf140 TBC1 domain family member 20 TBC1 domain family member 20 TBC1D20 WARBM4; TBC1 domain family member 20; TBC1 domain family, member 20
Gene Aliases: C20orf140; TBC1D20; WARBM4
UniProt ID: (Human) Q96BZ9
Entrez Gene ID: (Human) 128637
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.