Invitrogen
Manufacturer Code:PA5113235
Catalog # PIPA5113235
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human TBC1 domain family member 20 protein (1-237AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf140 TBC1 domain family member 20 TBC1 domain family member 20 TBC1D20 WARBM4; TBC1 domain family member 20; TBC1 domain family, member 20
Gene Aliases: 1110028I04Rik; 2810442O16Rik; AI414693; C20orf140; TBC1D20; WARBM4
UniProt ID: (Human) Q96BZ9, (Mouse) Q9D9I4
Entrez Gene ID: (Human) 128637, (Mouse) 67231
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.