OriGene
Manufacturer Code:CF810800
Catalog # 50-167-0316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5A4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 45-248 of human TAZ produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BTHS CMD3A EFE EFE2 G4.5 LVNCX Taz1; Protein G4.5; Tafazzin
Gene Aliases: BTHS; CMD3A; EFE; EFE2; G4.5; LVNCX; TAZ; Taz1
UniProt ID: (Human) Q16635
Entrez Gene ID: (Human) 6901
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.