Invitrogen
Manufacturer Code:MA520076
Catalog # PIMA520076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B3 |
Immunogen | TAZ (AAH11515 1 a.a. &bsim 262 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Barth syndrome); cardiomyopathy, dilated 3A (X-linked); endocardial fibroelastosis 2; Protein G4.5; Tafazzin; tafazzin (cardiomyopathy, dilated 3A (X-linked); tafazzin (cardiomyopathy, dilated 3A (X-linked); endocardial fibroelastosis 2; Barth syndrome); TAZ tafazzin (cardiomyopathy dilated 3A (X-linked) endocardial fibroelastosis 2 Barth syndrome) HGNC:11577 BTHS CMD3A EFE EFE2 G4.5 XAP-2 OTTHUMP00000061673 cardiomyopathy dilated 3A (X-linked) tafazzin XX-FW83563B9.3 FLJ27390 Taz1 LVNCX
Gene Aliases: BTHS; CMD3A; EFE; EFE2; G4.5; LVNCX; TAZ; Taz1
UniProt ID: (Human) Q16635
Entrez Gene ID: (Human) 6901, (Rat) 363521
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.