Invitrogen
Manufacturer Code:PA5114792
Catalog # PIPA5114792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAZ corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5031411C02Rik 9130012G04Rik AW107266 AW552613 Barth syndrome) BTHS cardiomyopathy dilated 3A (X-linked) CMD3A EFE EFE2 endocardial fibroelastosis 2 FLJ27390 G4.5 HGNC:11577 LVNCX OTTHUMP00000061673 protein G4.5 Tafazzin tafazzin (cardiomyopathy dilated 3A (X-linked) tafazzin (cardiomyopathy dilated 3A (X-linked) endocardial fibroelastosis 2 Barth syndrome) TAZ Taz1 XAP-2 XX-FW83563B9.3; Barth syndrome); cardiomyopathy, dilated 3A (X-linked); endocardial fibroelastosis 2; Protein G4.5; Tafazzin; tafazzin (cardiomyopathy, dilated 3A (X-linked); tafazzin (cardiomyopathy, dilated 3A (X-linked); endocardial fibroelastosis 2; Barth syndrome)
Gene Aliases: 5031411C02Rik; 9130012G04Rik; AW107266; AW552613; BTHS; CMD3A; EFE; EFE2; G4.5; LVNCX; TAZ; Taz1
UniProt ID: (Human) Q16635
Entrez Gene ID: (Human) 6901, (Rat) 363521, (Mouse) 66826
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.