Invitrogen
Manufacturer Code:PA537460
Catalog # PIPA537460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 570-620 of human TAUT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC10619 MGC131729 OTTHUMP00000208177 OTTHUMP00000208180 SC6A6 SLC6A6 Sodium- and chloride-dependent taurine transporter Solute carrier family 6 (neurotransmitter transporter taurine) member 6 Solute carrier family 6 member 6 TAUT; Sodium- and chloride-dependent taurine transporter; Solute carrier 6 ,member 6 (taurine transporter); solute carrier family 6 (neurotransmitter transporter, taurine), member 6; Solute carrier family 6 member 6; solute carrier family 6, member 6; taurine/beta-alanine transporter
Gene Aliases: AA589629; C80501; SLC6A6; TAUT
UniProt ID: (Human) P31641, (Mouse) O35316, (Rat) P31643
Entrez Gene ID: (Human) 6533, (Mouse) 21366, (Rat) 29464
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.