Invitrogen
Manufacturer Code:MA512805
Catalog # PIMA512805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TAU-5 |
Immunogen | Purified bovine microtubule-associated proteins |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: G protein beta1/gamma2 subunit-interacting factor 1; G protein beta1/gamma2 subunit-interacting factor 1 PHF-tau microtubule-associated protein tau isoform 4 neurofibrillary tangle protein paired helical filament-tau Tau5; Microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; Neurofibrillary tangle protein; Paired helical filament-tau; PHF-tau; protein phosphatase 1, regulatory subunit 103; Tau microtubule-associated protein
Gene Aliases: AI413597; AW045860; DDPAC; FTDP-17; MAPT; MAPTL; MSTD; Mtapt; MTBT1; MTBT2; PPND; PPP1R103; pTau; RNPTAU; TAU
UniProt ID: (Bovine) P29172, (Human) P10636, (Mouse) P10637
Entrez Gene ID: (Bovine) 281296, (Human) 4137, (Sheep) 101121024, (Rat) 29477, (Mouse) 17762
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.